Assay Details
| Gene Symbol: SERPINB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171288.1 | NP_001164759.1 | 5 - 6 | 714 | 76 | |
| XM_008261390.2 | XP_008259612.1 | 7 - 8 | 944 | 76 | ||
| XM_008261391.2 | XP_008259613.1 | 7 - 8 | 919 | 76 | ||
| XM_008261392.2 | XP_008259614.1 | 7 - 8 | 915 | 76 | ||
| XM_008261393.2 | XP_008259615.1 | 7 - 8 | 891 | 76 | ||
| XM_008261394.2 | XP_008259616.1 | 6 - 7 | 1399 | 76 | ||
| XM_017343656.1 | XP_017199145.1 | 7 - 8 | 1387 | 76 | ||
| XM_017343657.1 | XP_017199146.1 | 7 - 8 | 1362 | 76 | ||
More Information
Set Membership: |
|

SERPINB8
Genomic Map