Assay Details
Gene Symbol: PSMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171305.1 | NP_001164776.1 | 3 - 4 | 500 | 56 | |
XM_008264042.2 | XP_008262264.1 | 3 - 4 | 504 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
