Assay Details
| Gene Symbol: RBM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171335.1 | NP_001164806.1 | 10 - 11 | 1099 | 66 | |
| XM_008255967.2 | XP_008254189.1 | 11 - 12 | 1426 | 66 | ||
| XM_008255968.2 | XP_008254190.1 | 11 - 12 | 1426 | 66 | ||
| XM_008255969.2 | XP_008254191.1 | 10 - 11 | 1360 | 66 | ||
| XM_008255970.2 | XP_008254192.1 | 10 - 11 | 1360 | 66 | ||
| XM_017341621.1 | XP_017197110.1 | 11 - 12 | 1426 | 66 | ||
More Information
Set Membership: |
|

RBM39
Genomic Map