Assay Details
| Gene Symbol: RHBDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171412.1 | NP_001164883.1 | 1561 | 69 | ||
| XM_008252784.2 | XP_008251006.1 | 3444 | 69 | |||
| XM_017340021.1 | XP_017195510.1 | 3553 | 69 | |||
| XM_017340022.1 | XP_017195511.1 | 3483 | 69 | |||
| XM_017340023.1 | XP_017195512.1 | 3555 | 69 | |||
More Information
Set Membership: |
|

RHBDF1
Genomic Map