Assay Details
Gene Symbol: OLFR649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171470.1 | NP_001164941.1 | 1 | 453 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OLFR649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171470.1 | NP_001164941.1 | 1 | 453 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |