Assay Details
| Gene Symbol: RFX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171491.1 | NP_001164962.1 | 1 - 2 | 120 | 60 | |
| XM_008264046.2 | XP_008262268.1 | 2 - 3 | 285 | 60 | ||
| XM_008264047.2 | XP_008262269.1 | 3 - 4 | 209 | 60 | ||
| XM_008264049.2 | XP_008262271.1 | 2 - 3 | 293 | 60 | ||
| XM_017345518.1 | XP_017201007.1 | 2 - 3 | 281 | 60 | ||
| XM_017345519.1 | XP_017201008.1 | 2 - 3 | 279 | 60 | ||
More Information
Set Membership: |
|

RFX5
Genomic Map