Assay Details
| Gene Symbol: KCNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198982.1 | NP_001185911.1 | 1 | 1351 | 60 | |
| XM_017343247.1 | XP_017198736.1 | 2 | 3063 | 60 | ||
More Information
Set Membership: |
|

KCNA1
Genomic Map