Assay Details
Gene Symbol: TICAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204346.1 | NP_001191275.1 | 1 | 455 | 65 | |
XM_008254897.2 | XP_008253119.1 | 2 | 635 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
