Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204806.1 | NP_001191735.1 | 1 - 2 | 82 | 86 | |
| XM_017345526.1 | XP_017201015.1 | 1 - 2 | 346 | 86 | ||
More Information
Set Membership: |
|

FPGT
Genomic Map