Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204806.1 | NP_001191735.1 | 3 - 4 | 342 | 93 | |
| XM_017345527.1 | XP_017201016.1 | 3 - 4 | 354 | 93 | ||
More Information
Set Membership: |
|

FPGT
Genomic Map