Assay Details
Gene Symbol: EMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710733.3 | XP_002710779.1 | 5 - 6 | 410 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
