Assay Details
| Gene Symbol: RPL35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205225.1 | NP_001192154.1 | 1 | 28 | 77 | |
| GenBank mRNA | CU464532.1 | - | 1 | 36 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001205225.1 transcripts.
Set Membership: |
|

RPL35A
Genomic Map