Assay Details
| Gene Symbol: AKR1B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267588.1 | NP_001254517.1 | 3 - 4 | 401 | 102 | |
| GenBank mRNA | AB724112.1 | - | 3 - 4 | 401 | 102 | |
More Information
Set Membership: |
|

AKR1B10
Genomic Map