Assay Details
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281863.1 | NP_001268792.1 | 4 - 5 | 462 | 63 | |
XM_008270668.2 | XP_008268890.1 | 5 - 6 | 584 | 63 | ||
XM_017348812.1 | XP_017204301.1 | 5 - 6 | 469 | 63 | ||
XM_017348813.1 | XP_017204302.1 | 4 - 5 | 405 | 63 | ||
XM_017348814.1 | XP_017204303.1 | 3 - 4 | 244 | 63 | ||
GenBank mRNA | EB373533.1 | - | 1 - 2 | 107 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
