Assay Details
| Gene Symbol: AOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297488.1 | NP_001284417.1 | 16 - 17 | 1729 | 58 | |
| XM_017342642.1 | XP_017198131.1 | 16 - 17 | 1873 | 58 | ||
More Information
Set Membership: |
|

AOX2
Genomic Map