Assay Details
Gene Symbol: MBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713482.3 | XP_002713528.2 | 1676 | 63 | ||
XM_017344180.1 | XP_017199669.1 | 1676 | 63 | |||
XM_017344181.1 | XP_017199670.1 | 1676 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
