Assay Details
Gene Symbol: RAB6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002708705.3 | XP_002708751.1 | 7 - 8 | 1037 | 80 | |
XM_008264212.2 | XP_008262434.1 | 7 - 8 | 549 | 80 | ||
XM_017344791.1 | XP_017200280.1 | 7 - 8 | 1053 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
