Assay Details
Gene Symbol: LOC100342335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008273836.2 | XP_008272058.1 | 2207 | 165 | ||
XM_008273837.2 | XP_008272059.1 | 2149 | 165 | |||
XM_008273838.2 | XP_008272060.1 | 2191 | 165 | |||
XM_017350278.1 | XP_017205767.1 | 2560 | 165 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
