Assay Details
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008270701.2 | XP_008268923.1 | 2 - 3 | 249 | 126 | |
XM_008270702.2 | XP_008268924.1 | 3 - 4 | 439 | 126 | ||
XM_017348924.1 | XP_017204413.1 | 2 - 3 | 192 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
