Assay Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721884.3 | XP_002721930.2 | 143 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002721884.3 transcripts.
Set Membership: |
|

ZNF394
Genomic Map