Assay Details
| Gene Symbol: LOC100353349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343969.1 | XP_017199458.1 | 5 - 6 | 772 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100353349| Gene Symbol: LOC100353349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343969.1 | XP_017199458.1 | 5 - 6 | 772 | 64 | |
Set Membership: |
|