Assay Details
| Gene Symbol: TRIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710045.3 | XP_002710091.1 | 2 - 3 | 1314 | 66 | |
| XM_017340892.1 | XP_017196381.1 | 2 - 3 | 410 | 66 | ||
More Information
Set Membership: |
|

TRIB2
Genomic Map