Assay Details
| Gene Symbol: PIGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272370.2 | XP_008270592.1 | 6 | 3527 | 118 | |
| XM_008272371.2 | XP_008270593.1 | 6 | 4859 | 118 | ||
| XM_008272372.2 | XP_008270594.1 | 7 | 3639 | 118 | ||
| XM_008272374.2 | XP_008270596.2 | 5 | 2811 | 118 | ||
More Information
Set Membership: |
|

PIGA
Genomic Map