Assay Details
Gene Symbol: C14H3orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716273.3 | XP_002716319.2 | 3 - 4 | 409 | 108 | |
XM_017346865.1 | XP_017202354.1 | 3 - 4 | 586 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
