Assay Details
| Gene Symbol: UROD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715141.3 | XP_002715187.1 | 5 - 6 | 563 | 67 | |
| XM_008265351.2 | XP_008263573.1 | 5 - 6 | 760 | 67 | ||
More Information
Set Membership: |
|

UROD
Genomic Map