Assay Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720620.3 | XP_002720666.3 | 995 | 54 | ||
XM_008273589.2 | XP_008271811.2 | 995 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
