Assay Details
| Gene Symbol: METTL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719463.3 | XP_002719509.1 | 7 - 8 | 923 | 72 | |
| XM_008271665.2 | XP_008269887.1 | 7 - 8 | 926 | 72 | ||
| XM_008271666.2 | XP_008269888.1 | 7 - 8 | 925 | 72 | ||
| XM_008271667.2 | XP_008269889.1 | 7 - 8 | 925 | 72 | ||
More Information
Set Membership: |
|

METTL2A
Genomic Map