Assay Details
| Gene Symbol: ACOT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721235.2 | XP_002721281.1 | 541 | 60 | ||
| XM_017350736.1 | XP_017206225.1 | 541 | 60 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACOT8| Gene Symbol: ACOT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721235.2 | XP_002721281.1 | 541 | 60 | ||
| XM_017350736.1 | XP_017206225.1 | 541 | 60 | |||
Set Membership: |
|