Assay Details
| Gene Symbol: LOC100351754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262693.2 | XP_008260915.1 | 16 - 17 | 2008 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100351754| Gene Symbol: LOC100351754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262693.2 | XP_008260915.1 | 16 - 17 | 2008 | 91 | |
Set Membership: |
|