Assay Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713490.3 | XP_002713536.1 | 4 - 5 | 590 | 91 | |
| XM_002713491.3 | XP_002713537.1 | 4 - 5 | 590 | 91 | ||
| XM_017344099.1 | XP_017199588.1 | 4 - 5 | 590 | 91 | ||
More Information
Set Membership: |
|

RNF138
Genomic Map