Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266713.2 | XP_008264935.1 | 2 - 3 | 772 | 93 | |
| XM_017347013.1 | XP_017202502.1 | 2 - 3 | 223 | 93 | ||
More Information
Set Membership: |
|

CEP19
Genomic Map