Assay Details
| Gene Symbol: LPAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272961.2 | XP_008271183.1 | 3 - 4 | 574 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008272961.2 transcripts.
Set Membership: |
|

LPAR4
Genomic Map