Assay Details
Gene Symbol: LOC100343179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008250636.2 | XP_008248858.1 | 1295 | 87 | ||
XM_008250637.2 | XP_008248859.1 | 1129 | 87 | |||
XM_008250638.2 | XP_008248860.1 | 1010 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
