Assay Details
| Gene Symbol: GALE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715981.3 | XP_002716027.2 | 3 - 4 | 905 | 59 | |
| XM_017346469.1 | XP_017201958.1 | 3 - 4 | 413 | 59 | ||
More Information
Set Membership: |
|

GALE
Genomic Map