Assay Details
| Gene Symbol: LOC100344750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267611.2 | XP_008265833.1 | 1 - 2 | 124 | 82 | |
| XM_017347409.1 | XP_017202898.1 | 1 - 2 | 124 | 82 | ||
| XM_017347410.1 | XP_017202899.1 | 1 - 2 | 126 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008267611.2, XM_017347409.1, XM_017347410.1 transcripts.
Set Membership: |
|

LOC100344750
Genomic Map