Assay Details
| Gene Symbol: LOC100344750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267611.2 | XP_008265833.1 | 23 - 24 | 2873 | 86 | |
| XM_008267612.2 | XP_008265834.1 | 23 - 24 | 2770 | 86 | ||
| XM_017347409.1 | XP_017202898.1 | 19 - 20 | 2267 | 86 | ||
More Information
Set Membership: |
|

LOC100344750
Genomic Map