Assay Details
| Gene Symbol: LOC100344750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267611.2 | XP_008265833.1 | 8 - 9 | 1122 | 68 | |
| XM_008267612.2 | XP_008265834.1 | 8 - 9 | 1019 | 68 | ||
| XM_017347410.1 | XP_017202899.1 | 8 - 9 | 1124 | 68 | ||
More Information
Set Membership: |
|

LOC100344750
Genomic Map