Assay Details
Gene Symbol: PIGO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008270909.2 | XP_008269131.1 | 4 - 5 | 1061 | 63 | |
XM_008271099.2 | XP_008269321.1 | 4 - 5 | 1063 | 63 | ||
XM_017340409.1 | XP_017195898.1 | 4 - 5 | 1063 | 63 | ||
XM_017340427.1 | XP_017195916.1 | 3 - 4 | 743 | 63 | ||
XM_017340442.1 | XP_017195931.1 | 3 - 4 | 710 | 63 | ||
XM_017340464.1 | XP_017195953.1 | 3 - 4 | 971 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
