Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717696.3 | XP_002717742.1 | 7 - 8 | 1523 | 63 | |
| XM_008268734.2 | XP_008266956.1 | 7 - 8 | 1313 | 63 | ||
| XM_008268735.2 | XP_008266957.1 | 7 - 8 | 1346 | 63 | ||
| XM_017347891.1 | XP_017203380.1 | 5 - 6 | 873 | 63 | ||
More Information
Set Membership: |
|

BRINP3
Genomic Map