Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720586.3 | XP_002720632.1 | 1642 | 74 | ||
XM_008273553.2 | XP_008271775.1 | 1410 | 74 | |||
XM_008273554.2 | XP_008271776.1 | 2042 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
