Assay Details
Gene Symbol: STIMATE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713252.3 | XP_002713298.1 | 2 - 3 | 289 | 83 | |
XM_008260802.2 | XP_008259024.1 | 2 - 3 | 289 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
