Assay Details
| Gene Symbol: PRCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263350.2 | XP_008261572.1 | 2 - 3 | 289 | 78 | |
| XM_017344368.1 | XP_017199857.1 | 2 - 3 | 299 | 78 | ||
More Information
Set Membership: |
|

PRCP
Genomic Map