Assay Details
Gene Symbol: LOC100358417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008254993.2 | XP_008253215.2 | 1325 | 60 | ||
XM_017341035.1 | XP_017196524.1 | 1325 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
