Assay Details
| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717497.3 | XP_002717543.1 | 3 - 4 | 1341 | 76 | |
| XM_008268400.2 | XP_008266622.1 | 3 - 4 | 1360 | 76 | ||
| XM_017347774.1 | XP_017203263.1 | 3 - 4 | 1778 | 76 | ||
More Information
Set Membership: |
|

DUSP10
Genomic Map