Assay Details
| Gene Symbol: EGFL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262623.2 | XP_008260845.1 | 6 - 7 | 1542 | 61 | |
| XM_008262624.2 | XP_008260846.1 | 6 - 7 | 1542 | 61 | ||
| XM_017344990.1 | XP_017200479.1 | 6 - 7 | 1542 | 61 | ||
More Information
Set Membership: |
|

EGFL8
Genomic Map