Assay Details
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720440.3 | XP_002720486.1 | 709 | 86 | ||
XM_008273306.2 | XP_008271528.1 | 792 | 86 | |||
XM_008273307.2 | XP_008271529.1 | 771 | 86 | |||
XM_008273308.2 | XP_008271530.1 | 747 | 86 | |||
XM_008273309.2 | XP_008271531.1 | 631 | 86 | |||
XM_017350095.1 | XP_017205584.1 | 640 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
