Assay Details
Gene Symbol: IPCEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008263713.2 | XP_008261935.1 | 11 - 12 | 1231 | 70 | |
XM_008263715.2 | XP_008261937.1 | 11 - 12 | 1228 | 70 | ||
XM_008263717.2 | XP_008261939.1 | 7 - 8 | 1058 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
