Assay Details
Gene Symbol: C10H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008261548.2 | XP_008259770.1 | 9 - 10 | 1296 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C10H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008261548.2 | XP_008259770.1 | 9 - 10 | 1296 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |