Assay Details
| Gene Symbol: FUS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248700.2 | XP_008246922.1 | 439 | 58 | ||
| XM_017337771.1 | XP_017193260.1 | 436 | 58 | |||
| XM_017337772.1 | XP_017193261.1 | 439 | 58 | |||
| XM_017337773.1 | XP_017193262.1 | 439 | 58 | |||
More Information
Set Membership: |
|

FUS
Genomic Map