Assay Details
Gene Symbol: PRR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722306.3 | XP_002722352.1 | 865 | 108 | ||
XM_017338235.1 | XP_017193724.1 | 720 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
